MAT047030285 Matrix Scientific

5-Chloro-2-fluorobenzylamine

  • MDL number MFCD01631440
  • Cas Number : 261723-26-6
  • Molecular Weight : 159.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002099.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/002099.html
Availability details for
Quantity
  • Availability
  • Add to Cart