MAT047030288 Matrix Scientific

6-(2-Phenyl-ethenesulfonylamino)-hexanoic acid

  • MDL number MFCD00539985
  • Cas Number : 315692-14-9
  • Molecular Weight : 297.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011311.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011311.html
Availability details for
Quantity
  • Availability
  • Add to Cart