MAT047030296 Matrix Scientific

5-Chloro-2-fluorobenzyl bromide

  • MDL number MFCD01631437
  • Cas Number : 71916-91-1
  • Molecular Weight : 223.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.65
Country of Origin USA
Boiling point 44-45�5mm
Product Link http://www.matrixscientific.com/002100.html

Safety Information

hcodes Corrosive/Refrigerate/Lachrymator
MSDS http://www.matrixscientific.com/002100.html
Availability details for
Quantity
  • Availability
  • Add to Cart