MAT047030303 Matrix Scientific

4-Amino-3-hydroxytetrahydrothiophene,1,1-dioxide

  • MDL number MFCD00456619
  • Cas Number : 55261-00-2
  • Molecular Weight : 151.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192
Country of Origin USA
Product Link http://www.matrixscientific.com/011325.html

Safety Information

MSDS http://www.matrixscientific.com/011325.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart