MAT047030308 Matrix Scientific

4-Amino-benzoic acid isopropyl ester

  • MDL number MFCD00017108
  • Cas Number : 18144-43-9
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86
Country of Origin USA
Boiling point 190-192�5 Torr
Product Link http://www.matrixscientific.com/011329.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011329.html
Availability details for
Quantity
  • Availability
  • Add to Cart