MAT047030325 Matrix Scientific

4-Phenylethynyl-benzaldehyde

  • MDL number MFCD00114585
  • Cas Number : 57341-98-7
  • Molecular Weight : 206.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102
Country of Origin USA
Product Link http://www.matrixscientific.com/011345.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011345.html
Availability details for
Quantity
  • Availability
  • Add to Cart