MAT047030352 Matrix Scientific

3-Chloro-2-fluorocinnamic acid

  • MDL number MFCD01631392
  • Cas Number : 261762-62-3
  • Molecular Weight : 200.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/002105.html

Safety Information

hcodes Irritant-Harmful
MSDS http://www.matrixscientific.com/002105.html
Availability details for
Quantity
  • Availability
  • Add to Cart