MAT047030389 Matrix Scientific

3-Hydroxymethyl-4-methoxy-benzaldehyde

  • MDL number MFCD00778411
  • Cas Number : 76646-42-9
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50
Country of Origin USA
Product Link http://www.matrixscientific.com/011402.html

Safety Information

MSDS http://www.matrixscientific.com/011402.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart