MAT047030390 Matrix Scientific

1-(4-Ethylphenyl)-2-oxopyrrolidine-4-carboxylic acid

  • MDL number MFCD00469851
  • Cas Number : 133748-26-2
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011403.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011403.html
Availability details for
Quantity
  • Availability
  • Add to Cart