MAT047030428 Matrix Scientific

3-(1-Carboxyethyl)thiazolidine-4-one-2-thione

  • MDL number MFCD00457538
  • Cas Number : 39137-13-8
  • Molecular Weight : 205.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-144
Country of Origin USA
Product Link http://www.matrixscientific.com/011438.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011438.html
Availability details for
Quantity
  • Availability
  • Add to Cart