MAT047030433 Matrix Scientific

2-Amino-5-(2-thienyl)-1,3,4-thiadiazole

  • MDL number MFCD01043763
  • Cas Number : 4683-00-5
  • Molecular Weight : 183.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-207
Country of Origin USA
Product Link http://www.matrixscientific.com/011442.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011442.html
Availability details for
Quantity
  • Availability
  • Add to Cart