MAT047030441 Matrix Scientific

5-Chloro-2-fluorophenylacetic acid

  • MDL number MFCD01631439
  • Cas Number : 261762-97-4
  • Molecular Weight : 188.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-118
Country of Origin USA
Product Link http://www.matrixscientific.com/002113.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002113.html
Availability details for
Quantity
  • Availability
  • Add to Cart