MAT047030463 Matrix Scientific

5-Chloro-2-fluorophenylacetonitrile

  • MDL number MFCD01631438
  • Cas Number : 75279-54-8
  • Molecular Weight : 169.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002115.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/002115.html
Availability details for
Quantity
  • Availability
  • Add to Cart