MAT047030493 Matrix Scientific

1-Allyl-1-phenylbut-3-enylamine

  • MDL number MFCD01156616
  • Cas Number : 19243-52-8
  • Molecular Weight : 187.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 87-90�75 Torr
Product Link http://www.matrixscientific.com/011496.html

Safety Information

MSDS http://www.matrixscientific.com/011496.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart