MAT047030503 Matrix Scientific

4-(3,5-Dimethyl-1H-pyrazol-1-yl)-6-methylpyrimidin-2-amine

  • MDL number MFCD01175033
  • Cas Number : 91716-38-0
  • Molecular Weight : 203.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-116
Country of Origin USA
Product Link http://www.matrixscientific.com/011505.html

Safety Information

MSDS http://www.matrixscientific.com/011505.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart