MAT047030526 Matrix Scientific

8-Methoxy-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinoline-4-carboxylic acid

  • MDL number MFCD01539374
  • Cas Number : 247225-89-4
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011526.html

Safety Information

MSDS http://www.matrixscientific.com/011526.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart