MAT047030554 Matrix Scientific

(5,6,7,8-Tetrahydro-benzo[4,5]thieno-[2,3-d]pyrimidin-4-ylamino)-acetic acid

  • MDL number MFCD00438867
  • Cas Number : 313534-22-4
  • Molecular Weight : 263.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011549.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011549.html
Availability details for
Quantity
  • Availability
  • Add to Cart