MAT047030556 Matrix Scientific

2,6-Bis-(2-methyl-allyl)-1,2,3,6-tetrahydro-pyridine

  • MDL number MFCD01936795
  • Cas Number : 439125-79-8
  • Molecular Weight : 191.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011551.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011551.html
Availability details for
Quantity
  • Availability
  • Add to Cart