MAT047030604 Matrix Scientific

(4-Methoxy-benzyl)-(2-methoxy-1-methyl-ethyl)-amine

  • MDL number MFCD00981067
  • Cas Number : 353777-78-3
  • Molecular Weight : 209.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011595.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011595.html
Availability details for
Quantity
  • Availability
  • Add to Cart