MAT047030716 Matrix Scientific

4-[(3-Methylpyridin-2-yl)amino]-4-oxobutanoic acid

  • MDL number MFCD01005032
  • Cas Number : 200417-51-2
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-180
Country of Origin USA
Product Link http://www.matrixscientific.com/011693.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011693.html
Availability details for
Quantity
  • Availability
  • Add to Cart