MAT047030809 Matrix Scientific

6,8-Dichloro-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinoline-4-carboxylic acid

  • MDL number MFCD01122992
  • Cas Number : 470693-57-3
  • Molecular Weight : 284.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011777.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011777.html
Availability details for
Quantity
  • Availability
  • Add to Cart