MAT047030821 Matrix Scientific

2-Methyl-5-morpholin-4-ylmethyl-furan-3-carboxylic acid

  • MDL number MFCD01797038
  • Cas Number : 309271-37-2
  • Molecular Weight : 225.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011788.html

Safety Information

MSDS http://www.matrixscientific.com/011788.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart