MAT047030840 Matrix Scientific

6-Ethyl-thieno[2,3-b]quinoline-2-carboxylic acid

  • MDL number MFCD02588425
  • Cas Number : 462066-95-1
  • Molecular Weight : 257.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011805.html

Safety Information

MSDS http://www.matrixscientific.com/011805.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart