MAT047030853 Matrix Scientific

N-(4-Methoxyphenyl)-N-(methylsulfonyl)glycine

  • MDL number MFCD02219785
  • Cas Number : 425616-70-2
  • Molecular Weight : 259.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011817.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011817.html
Availability details for
Quantity
  • Availability
  • Add to Cart