MAT047030865 Matrix Scientific

3-Acetyl-2-methylquinoline-4-carboxylic acid

  • MDL number MFCD00957757
  • Cas Number : 106380-95-4
  • Molecular Weight : 229.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198
Country of Origin USA
Product Link http://www.matrixscientific.com/011828.html

Safety Information

MSDS http://www.matrixscientific.com/011828.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart