MAT047030866 Matrix Scientific

4-Fluoro-2-methylanisole

  • MDL number MFCD00143319
  • Cas Number : 399-54-2
  • Molecular Weight : 140.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 69-70�m
Product Link http://www.matrixscientific.com/002156.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002156.html
Availability details for
Quantity
  • Availability
  • Add to Cart