MAT047030911 Matrix Scientific

5-Fluoro-2-methylbenzoyl chloride

  • MDL number MFCD00236681
  • Cas Number : 21900-39-0
  • Molecular Weight : 172.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002161.html

Safety Information

hcodes Corrosive/Refrigerate
MSDS http://www.matrixscientific.com/002161.html
Availability details for
Quantity
  • Availability
  • Add to Cart