MAT047030924 Matrix Scientific

2-Chloro-6-thien-2-ylnicotinic acid

  • MDL number MFCD02232164
  • Cas Number : 462066-77-9
  • Molecular Weight : 239.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011880.html

Safety Information

MSDS http://www.matrixscientific.com/011880.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart