MAT047030962 Matrix Scientific

5-Benzooxazol-2-yl-2-methyl-phenylamine

  • MDL number MFCD00579038
  • Cas Number : 292644-38-3
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011915.html

Safety Information

MSDS http://www.matrixscientific.com/011915.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart