MAT047030964 Matrix Scientific

2-Methyl-5-(5-methyl-benzooxazol-2-yl)-phenylamine

  • MDL number MFCD00579065
  • Cas Number : 293737-74-3
  • Molecular Weight : 238.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011917.html

Safety Information

MSDS http://www.matrixscientific.com/011917.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart