MAT047030965 Matrix Scientific

2-Methyl-5-(6-methyl-benzooxazol-2-yl)-phenylamine

  • MDL number MFCD00579039
  • Cas Number : 293737-75-4
  • Molecular Weight : 238.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011918.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011918.html
Availability details for
Quantity
  • Availability
  • Add to Cart