MAT047030969 Matrix Scientific

2-(3,4-Dimethoxy-phenyl)-benzooxazol-5-ylamine

  • MDL number MFCD02069646
  • Cas Number : 389081-33-8
  • Molecular Weight : 270.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011921.html

Safety Information

MSDS http://www.matrixscientific.com/011921.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart