MAT047031011 Matrix Scientific

3-Chloro-2-methylbenzylamine

  • MDL number MFCD00053644
  • Cas Number : 226565-61-3
  • Molecular Weight : 155.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.14
Country of Origin USA
Product Link http://www.matrixscientific.com/002171.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002171.html
Availability details for
Quantity
  • Availability
  • Add to Cart