MAT047031026 Matrix Scientific

4-(3-Formyl-2,5-dimethyl-pyrrol-1-yl)-3-methyl-benzoic acid

  • MDL number MFCD02629509
  • Cas Number : 409353-44-2
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011972.html

Safety Information

MSDS http://www.matrixscientific.com/011972.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart