MAT047031032 Matrix Scientific

N-(3-Amino-phenyl)-4-fluoro-benzenesulfonamide

  • MDL number MFCD02593904
  • Cas Number : 436089-66-6
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011978.html

Safety Information

MSDS http://www.matrixscientific.com/011978.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart