MAT047031038 Matrix Scientific

1-Allyl-1-p-tolyl-but-3-enylamine

  • MDL number MFCD02735476
  • Cas Number : 189167-67-7
  • Molecular Weight : 201.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011983.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011983.html
Availability details for
Quantity
  • Availability
  • Add to Cart