MAT047031044 Matrix Scientific

3-Chloro-2-methylbenzonitrile

  • MDL number MFCD00045598
  • Cas Number : 54454-12-5
  • Molecular Weight : 151.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 21-23
Purity 98
Country of Origin USA
Boiling point 120�mm
Product Link http://www.matrixscientific.com/002175.html

Safety Information

hcodes Irritant-Harmful
MSDS http://www.matrixscientific.com/002175.html
Availability details for
Quantity
  • Availability
  • Add to Cart