MAT047031068 Matrix Scientific

5-Oxo-1-phenyl-pyrrolidine-3-carboxylic acid

  • MDL number MFCD00207656
  • Cas Number : 39629-86-2
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189-190
Country of Origin USA
Product Link http://www.matrixscientific.com/012010.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012010.html
Availability details for
Quantity
  • Availability
  • Add to Cart