MAT047031079 Matrix Scientific

2-Chloro-5-pyrrol-1-yl-benzoic acid

  • MDL number MFCD03036852
  • Cas Number : 53242-68-5
  • Molecular Weight : 221.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-128
Country of Origin USA
Product Link http://www.matrixscientific.com/012020.html

Safety Information

MSDS http://www.matrixscientific.com/012020.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart