MAT047031101 Matrix Scientific

Quinoline-6-carbaldehyde

  • MDL number MFCD00805836
  • Cas Number : 4113-04-6
  • Molecular Weight : 157.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-76
Storage temp. 18899-64-4
Country of Origin USA
Product Link http://www.matrixscientific.com/012040.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/012040.html
Availability details for
Quantity
  • Availability
  • Add to Cart