MAT047031104 Matrix Scientific

3-Methoxy-4-(4-methyl-piperidine-1-sulfonyl)-phenylamine

  • MDL number MFCD02746180
  • Cas Number : 436091-53-1
  • Molecular Weight : 284.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012043.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012043.html
Availability details for
Quantity
  • Availability
  • Add to Cart