MAT047031118 Matrix Scientific

7-Bromo-1,3,4,10-tetrahydro-2H-acridin-9-one

  • MDL number MFCD02737904
  • Cas Number : 443662-39-3
  • Molecular Weight : 278.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >350
Country of Origin USA
Product Link http://www.matrixscientific.com/012055.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012055.html
Availability details for
Quantity
  • Availability
  • Add to Cart