MAT047031121 Matrix Scientific

5-tert-Butyl-4-hydroxymethyl-2-methyl-furan-3-carboxylic acid

  • MDL number MFCD02737949
  • Cas Number : 462068-43-5
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012058.html

Safety Information

MSDS http://www.matrixscientific.com/012058.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart