MAT047031123 Matrix Scientific

2-Chloro-5-(3,5-dimethyl-pyrazol-1-yl)-benzoic acid

  • MDL number MFCD03014629
  • Cas Number : 401827-60-9
  • Molecular Weight : 250.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012059.html

Safety Information

MSDS http://www.matrixscientific.com/012059.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart