MAT047031128 Matrix Scientific

2-(2-Oxo-pyrrolidin-1-yl)-propionic acid

  • MDL number MFCD05237228
  • Cas Number : 67118-32-5
  • Molecular Weight : 157.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012064.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012064.html
Availability details for
Quantity
  • Availability
  • Add to Cart