MAT047031133 Matrix Scientific

5-Fluoro-2-methylphenylacetic acid

  • MDL number MFCD01631530
  • Cas Number : 261951-75-1
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002183.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002183.html
Availability details for
Quantity
  • Availability
  • Add to Cart