MAT047031135 Matrix Scientific

3-(Pyridin-2-ylsulfamoyl)-benzoic acid

  • MDL number MFCD04113964
  • Cas Number : 683265-90-9
  • Molecular Weight : 278.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232-233
Country of Origin USA
Product Link http://www.matrixscientific.com/012070.html

Safety Information

MSDS http://www.matrixscientific.com/012070.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart