MAT047031141 Matrix Scientific

(1-Furan-2-yl-but-3-enyl)-thiophen-2-ylmethyl-amine

  • MDL number MFCD03075631
  • Cas Number : 799779-51-4
  • Molecular Weight : 233.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012076.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012076.html
Availability details for
Quantity
  • Availability
  • Add to Cart