MAT047031147 Matrix Scientific

5-(4-Ethoxy-phenyl)-7-methyl-pyrazolo-[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD01194454
  • Cas Number : 354542-40-8
  • Molecular Weight : 297.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012081.html

Safety Information

MSDS http://www.matrixscientific.com/012081.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart