MAT047031155 Matrix Scientific

4-Fluoro-2-methylphenyl isothiocyanate

  • MDL number MFCD00046805
  • Cas Number : 52317-97-2
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Boiling point 102�mm
Product Link http://www.matrixscientific.com/002185.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002185.html
Availability details for
Quantity
  • Availability
  • Add to Cart